missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polyethylene Stoppers

Product Code. 12997326
Click to view available options
Standard Taper Joint:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12997326

Brand: Azlon™ TRF597

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. Azlon™ Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.

Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.

  • Manufactured from polyethylene
  • Available in white and red
  • Suitable for use with both Azlon and glass volumetric ware
  • Enlarged head provides a good grip and protects glassware
  • Ribbed cone ensures a secure leak free seal
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Glass or Plastic volumetric flasks and measuring cylinders etc
Standard Taper Joint 29/32
Color White, Red
Material Polyethylene
Product Type Tapered Stopper
Shape Ribbed Cone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.