missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Terephthalate, (contains 30% glass particles as reinforcer), TRC

Product Code. 30426866
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426866 Supplier Toronto Research Chemicals Supplier No. TRCP688530100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Terephthalate (PET)
CAS 25038-59-9
Molecular Formula (C10 H8 O4)n
Quantity 100 mg
Synonym Poly(oxy-1,2-ethanediyloxycarbonyl-1,4-phenylenecarbonyl), Poly(oxyethyleneoxyterephthaloyl) (8CI), 1,4-Benzenedicarboxylic acid','dimethyl ester','polymer with 1,2-ethanediol, 1,4-Benzenedicarboxylic acid','polymer with 1,2-ethanediol, Dimethyl terephthalate-1,2-ethanediol polymer','SRU, Dimethyl terephthalate-ethylene carbonate copolymer','SRU, Dimethyl terephthalate-ethylene glycol block copolymer, Dimethyl terephthalate-ethylene glycol copolymer, Dimethyl terephthalate-ethylene glycol copolymer','SRU, Dimethyl terephthalate-ethylene glycol polymer','SRU, Dimethyl terephthalate-ethylene glycol-silica graft copolymer, Divinycell P 100, Divinyl terephthalate-ethylene glycol copolymer, Ethylene carbonate-terephthalic acid copolymer','SRU, Ethylene glycol-terephthalic acid copolymer, Ethylene glycol-terephthalic acid copolymer','SRU, Ethylene glycol-terephthalic acid polymer, Ethylene glycol-terephthalic acid polymer SRU, Ethylene glycol-terephthalic acid-triclosan graft copolymer, Ethylene glycol-terephthalic chloride copolymer','SRU, Ethylene terephthalate polymer, Monoethylene glycol-terephthalic acid copolymer SRU, Poly(diethyl terephthalate), Poly(ethylene glycol terephthalate), Poly(ethylene terephthalate), Poly(ethylene terephthalate)
SMILES [*]OCCOC(=O)c1ccc(cc1)C(=O)[*]
Molecular Weight (g/mol) 192.17
Formula Weight 192.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.