missing translation for 'onlineSavingsMsg'
Learn More

GUERIN PLASTIQUES Polyethylene Sleeves

Product Code. 11856543
Change view
Click to view available options
Size:
0.1 Thick x 100Wmm
0.1 Thick x 120Wmm
0.1 Thick x 140Wmm
0.1 Thick x 160Wmm
0.1 Thick x 180Wmm
0.1 Thick x 200Wmm
0.1 Thick x 250Wmm
0.1 Thick x 300Wmm
0.1 Thick x 350Wmm
0.1 Thick x 55Wmm
0.1 Thick x 64Wmm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11856543 0.1 Thick x 350Wmm Each
11836523 0.1 Thick x 160Wmm Each
11806523 0.1 Thick x 100Wmm Each
11856523 0.1 Thick x 200Wmm Each
11846543 0.1 Thick x 300Wmm Each
11866523 0.1 Thick x 250Wmm Each
11816523 0.1 Thick x 120Wmm Each
11826523 0.1 Thick x 140Wmm Each
11846523 0.1 Thick x 180Wmm Each
11816503 0.1 Thick x 55Wmm Each
11896513 0.1 Thick x 64Wmm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 11856543 Supplier GUERIN PLASTIQUES Supplier No. 50162
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    161.74 € / Pack of 1
    Save 28.26 €
    14% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.

  • 350m long in total
  • Available in a variety of widths

Specifications

Quantity 1/Pk.
Type Sleeve
Material Polyethylene
For Use With (Equipment) Food Quality
Length (Metric) 350 m
Size 0.1 Thick x 350Wmm
Color Transparent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.