missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polyethylene Tray/Sump Combination for Shelf #29935 or 4 Gallon Safety Cabinet

Product Code. 11945534
Change view
Click to view available options
Color:
White
Unit Size:
Each
Product Code. Color unitSize
11945534 White Each
1 options
This item is not returnable. View return policy

Product Code. 11945534

Brand: Justrite™ 25934

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase the efficiency and space of existing cabinetry. The Justrite™ Polyethylene Tray/Sump Combination for Shelf #29935 or 4 Gallon Safety Cabinet is a great way protect cabinets from any leaks/spills with a poly tray that can fit over the shelf, or inside the bottom sump.

Specifications

Color White
For Use With (Equipment) Fits shelf for 4-gallon cabinet
Dimensions (L x W) 337 x 346 mm (13.25 x 13.265 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.