missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene, UHMW

Product Code. 11384998
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
11384998 50 g 50g
11364998 250 g 250g
11374998 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11384998

Brand: Thermo Scientific Alfa Aesar 043951.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-88-4 | (C2H4)n | 28.05 g/mol

Polyethylene, UHMW fibers is used in armor, especially personal armor, vehicle armor, climbing equipments, fishing line, bow strings and cut-resistant gloves. Its sheet is used as synthetic ice in ice rinks . In medical field, it is considered as a vital biomaterial used in hip, knee, for spine implants. It is also used in the production of polyvinyl chloride windows and doors, hydraulic seals and bearings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-88-4
Molecular Formula (C2H4)n
Molecular Weight (g/mol) 28.05
MDL Number MFCD00084423
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
PubChem CID 6325
ChEBI CHEBI:18153
IUPAC Name ethene
SMILES *-CC-*

Specifications

Physical Form Powder
Solubility Information Swellable in high temperatures with chlorinated aromatics. Insoluble in water.
Quantity 50 g
Odor Odorless
Chemical Name or Material Polyethylene, UHMW

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.