missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Wash Bottle
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
70% Ethanol
Acetone
Custom Write-on
Ethyl Acetate
Machine Oil
Methyl Ethyl Ketone
Soap
Toluene
Water
Closure Color:
Blue
Green
Natural
Red
Unit Size:
Pack of 4
Descripción
- Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Color-coded polypropylene closures aid in quick and easy identification
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- 500ml - 4/bag, 1000ml - 4/bag; assortment is 6 per bag, includes one each: Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB
Note: Use LYOB only with solvents compatible with LDPE
Especificaciones
Especificaciones
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Translucent |
| Diameter (Metric) Outer Neck | 53mm |
| Includes | Blue Polypropylene Cap |
| Material | Polyethylene |
| Quantity | 4Pack |
| Sterility | Non-sterile |
| Closure Color | Blue |
| Certifications/Compliance | Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido