missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Wash Bottle

Product Code. p-8007859 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
70% Ethanol
Acetone
Custom Write-on
Ethyl Acetate
Machine Oil
Methyl Ethyl Ketone
Soap
Toluene
Water
Closure Color:
Blue
Green
Natural
Red
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 12388118

Brand: Bel-Art™ F117160016

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene. Bel Art™ Polyethylene Wide Mouth Wash Bottle feature a wide mouth 53mm (2 in.) for easy refilling.

  • Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Color-coded polypropylene closures aid in quick and easy identification
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • 500ml - 4/bag, 1000ml - 4/bag; assortment is 6 per bag, includes one each: Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB

Note: Use LYOB only with solvents compatible with LDPE

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Diameter (Metric) Outer Neck 53mm
Includes Red Polypropylene Cap
Material Polyethylene
Quantity 4Pack
Sterility Non-sterile
Closure Color Red
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
Chemical Label Toluene
Closure Material Polypropylene
Closure Type Red Polypropylene Cap
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.