missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, 50% Aqueous Solution, MP Biomedicals™

Product Code. 11495362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495362 100 mL 100mL
11405372 500 mL 500mL
11415372 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495362 Supplier MP Biomedicals Supplier No. 0219544480
  • ONLINE EXCLUSIVE PRICE
    76.40 €
    72.45 € / 100mL
    Save 3.95 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Physical Form Viscous Liquid
Quantity 100 mL
Formula Weight 50,000 to 100,000
Chemical Name or Material Polyethyleneimine
compliance-icons
Product Identifier
  • Polyethyleneimine, 50% aqueous solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: polyethyleneimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.