missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, branched, M.W. 1,200, 99%

Product Code. 15422397
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422397 500 g 500g
15402397 25 g 25g
15412397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422397 Supplier Thermo Scientific Alfa Aesar Supplier No. 045594.36
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Color Yellow
Physical Form Liquid
Solubility Information Soluble in hot water at a low pH,methanol,ethanol or chloroform. Insoluble in cold water,benzene,ethyl ether and acetone.
Quantity 500 g
Percent Purity 99%
Odor Amine-like
Chemical Name or Material Polyethyleneimine, Branched, M.W. 1,200
compliance-icons
Product Identifier
  • Polyethyleneimine, branched, M.W. 1,200
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polyethyleneimine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.