missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched

Product Code. 10592354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10592354 1 kg Glass bottle 1kg
10720881 100 g Glass bottle 100g
10499650 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10592354 Supplier Thermo Scientific Acros Supplier No. 178570010
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6 , 7732-18-5
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Physical Form Viscous Liquid
Boiling Point 100.0°C
Linear Formula (-CH2CH2NH-)x-(CH2CH2N(CH2CH2NH2)-)y
Density 1.0700g/mL
Solubility Information Solubility in water: soluble.
Quantity 1 kg
Flash Point >110°C
Percent Purity 48 to 52% (solids)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.07
Chemical Name or Material Polyethyleneimine, 50 wt. % Aqueous Solution, Average M.W. 50-60000
Show More Show Less
compliance-icons
Product Identifier
  • Polyethyleneimine, 50 wt% aqueous solution, approx. M.N. 60,000
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.