missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, MP Biomedicals

Product Code. 11485362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485362 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485362 Supplier MP Biomedicals Supplier No. 0219544450
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

9002-98-6 (Polymin P) 50% Aqueous Solution

Specifications

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
MDL Number MFCD00084427
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
Quantity 50 mL
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
SMILES *-CCNCCN(-*)CCN-*
IUPAC Name aziridine
Molecular Weight (g/mol) 43.07
PubChem CID 9033
ChEBI CHEBI:30969
Chemical Name or Material Polyethyleneimine, 50%, Aqueous Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Polyethyleneimine, 50% Aqueous Solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: polyethylene amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.