missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine on silica beads, anion exchange resin, benzylated, 20-40 mesh, Thermo Scientific™

Product Code. 11399448
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399448 50 mL 50mL
11309458 10 mL 10mL
11389448 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399448 Supplier Thermo Scientific Alfa Aesar Supplier No. 043304.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solid phase organic synthesis, affinity chromatography, catalyst support

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Physical Form Beads
Quantity 50 mL
Mesh Size 20 to 40 Mesh
Sensitivity Hygroscopic
Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin, Benzylated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.