missing translation for 'onlineSavingsMsg'
Learn More

Polyethylenimine, TRC

Product Code. 30003468
Change view
Click to view available options
Quantity:
25 mL
50 mL
100 mL
Unit Size:
100mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003468 50 mL 50mL
30003300 25 mL 25mL
30011793 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003468 Supplier Toronto Research Chemicals Supplier No. TRCP92697550ML
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6
Molecular Formula -HN-[CH 2 CH 2 NH-]n
Synonym Adcote 372, Adcote372MW;Aldrich 408719;Aldrich 482595;Aziridine Polymer;Basocoll PR 8086;Basomin G 500;Basonal White FO 1;Bufloc 595;CF 218;CF 218 (Polymer);CP 8994;Corcat P 100;Corcat P 12;Corcat P 145;Corcat P 150;Corcat P 18;Corcat P 200;Corcat P 600;Duramax 1007;EA 275;EAz 1300;EAz 800;EC 005;EL 402;EL 420;EP 108;EP 108 (Polymer);Emerlube 6717;Epomin;Ethyleneimine Homopolymer;Ethylenimine Homopolymer;Everamine;Everamine 150T;Everamine 210T;Everamine 500T;Everamine 50T;F 2S;FC;Fluka P 3142;G 100;G 20;G 20WF;G 35;G 500;HF;HG 20;HM 2000;HZ 20;HZ 20 (Polyamine);IP 232;JS 980;K 203C;Katax 6760;L 771;Liposol G;Loxanol MI 6730;Loxanol MI 6735;Lugalvan G 15;Lugalvan G 20;Lugalvan G 35;Lugalvan G 50;Lupasol 800;Lupasol P-WF;Lupasol PN 40;Lupasol PR;Lupasol PR 8515;Lupasol PS;Lupasol R;Lupasol SN;Lupasol WF;Lupasoli G;Lupazol;Luposal P;Luprasol SK;Mica A 131X;Montrek 1000;Montrek 12;Montrek 18;Montrek 6;Montrek 600;OEI 800;Oribain EL 420;P 0381;P 100;P 100 (Polyamine);P 1000;P 1030;P 200;P 3142;P 3143;P 600;P 600XE;P 70;P 70 (Polyamine);PC 8994;PEI;PEI 1;PEI 100;PEI 1000;PEI 1120;PEI 12;PEI 18;PEI 2;PEI 300;PEI 400;PEI 6;PEI 600;PEI 700000;PEO 113V;PR 20;PR 20 (Release Agent);PR 8515;Poly 8;Polyaziridine;Polyethenimide;Polyimin P;Polymin 6;Polymin FL;Polymin G 100;Polymin G 15M;Polymin G 20;Polymin G 35;Polymin G 500;Polymin HM;Polymin HS;Polymin PR 9711;Polymin PR 971L;Polymin SKA;Polymin SNA;Polymin WF;Polymin Waterfree;Rewin CLE;Rhenocure DR;SN;SP 003;SP 006;SP 012;SP 012D;SP 018;SP 1050;SP 110;SP 200;SP 300;SP 400;SP 400 (Polyimine);SP 600;Sedipur CL 930;T 13A;TS 280;TS 280 (Crosslinking Agent);Titabond 185E;Titabond T 100;Toyobain 210K;Toyobine 210K;Tydex 12;UN 3082;UP 300;UP 300 (Polyamine);WF;XA 1007;XUS 19036.01
SMILES NCCN(CCNCCN(CCN(CCNCCN)CCN(CCN)CCN)CCNCCN)CCN

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C2H5N/c1-2-3-1/h3H,1-2H2
Chemical Name or Material Polyethylenimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.