missing translation for 'onlineSavingsMsg'
Learn More

Polygalacturonic Acid (Technical Grade), TRC

Product Code. 30417189
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417189 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417189 Supplier Toronto Research Chemicals Supplier No. TRCP6891005G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25990-10-7
Molecular Formula (C6 H10 O7)n
Synonym Galacturonic acid polymer
IUPAC Name poly-(2S,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxo-hexanoic acid
SMILES O[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H10O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h1-5,8-11H,(H,12,13)/t2-,3+,4+,5-/m0/s1
Chemical Name or Material Polygalacturonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.