missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 300 to 12 C4

Brand:  Macherey-Nagel™ 711022.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10032801

  • 3425.00 € / 1000g
Estimated Shipment: 28-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10 to 15 μm
POLYGOPREP 300 to 12
1 kg
Polygoprep
300 Å
2 to 8
POLYGOPREP C4
Liquid Chromatography
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers