missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram Alox-100 UV254 Polyester Sheets

Brand:  Macherey-Nagel™ 807033

Code : 50

Additional Details : Weight : 0.01000kg

Product Code. 10194661

  • 243.00 € / Pack of 15
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

60 Å
Polyester Sheet
1 mm
15 Pk.
UV254
20 x 20 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers