Learn More
Macherey-Nagel™ Alox-25 UV254 TLC Glass Plates
Product Code. 10184661 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10184661 20 x 20 cm Pack of 25
10584611 5 x 20 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10184661 Supplier Macherey-Nagel™ Supplier No. 807023
Add to basket
Add to basket
Specifications
Pore Size 60 Å
Quantity 25 Pk.
Type Glass Plates
Fluorescent Indicator UV254
Thickness (Metric) 0.25 mm
Dimensions (L x W) 20 x 20 cm