missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram Alox N/UV254 Polyester Sheets

Product Code. 10252901 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
4 x 8 cm
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10252901 4 x 8 cm Pack of 50
10499171 20 x 20 cm Pack of 25
10236381 5 x 20 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10252901 Supplier Macherey-Nagel™ Supplier No. 802021
  • ONLINE EXCLUSIVE PRICE
    47.55 €
    36.79 € / Pack of 50
    Save 10.76 €
    22% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Macherey-Nagel Polygram Alox N/UV254 Polyester Sheets, Dimensions: 4 x 8 cm, Pore Size: 60 A, Fluorescent Indicator: UV254, Thickness: 0.2 mm

Specifications

Backing Material Polyester
Material Aluminum Oxide, Alox N UV254
Pore Size 60 Å
Quantity 50/Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Sorbent Aluminum Oxide, Alox N UV254
Thickness (Metric) 0.2 mm
Dimensions (L x W) 4 x 8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.