missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram CEL 300 Polyester Sheets

Product Code. 10137071 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
4 x 8 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10137071 20 x 20 cm Pack of 25
10304411 4 x 8 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10137071 Supplier Macherey-Nagel™ Supplier No. 801013
  • ONLINE EXCLUSIVE PRICE
    180.00 €
    138.96 € / Pack of 25
    Save 41.04 €
    22% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TLC polyester sheets with native fibrous cellulose. Fiber length (95 %) 2–20 μm, average degree of polymerization 400–500, specific surface acc. to Blaine 15000 cm2/g; ≤ 20 ppm Fe, 6 ppm Cu, 7 ppm P; dichloromethane extract ≤ 0.25 %; residue on ignition at 850 °C ≤ 1500 ppm.

TLC Polyester sheets, cellulose layer, POLYGRAM CEL 300

  • Base material: Polyester sheets coated in cellulose
  • Mode: Normal Phase
  • Layer Hardness: Standard, Soft

Adsorbent:

  • Polyester sheets coated in cellulose
  • Phase CEL 300
  • UV Indicator: No
  • Particle Type: Fully porous particles (FPP)
  • Particle Shape: Irregular
  • Particle size 2-20 μm
  • Binder: Yes

Indicators:

This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).

Binder:

Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Recommended Applications:

Amino acids, Carbohydrates, Carboxylic acids, Polar substances

 

General TLC Glass Plate Features from Macherey-Nagel:

  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
  • The plate can be easily labelled for clear identification
  • Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
  • Increased separation efficiency due to an optimized particle size distribution
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber

Specifications

Quantity 25 Pk.
Type Polyester Sheet
Thickness (Metric) 0.1 mm
Dimensions (L x W) 20 x 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.