Learn More
Macherey-Nagel™ Polygram CEL 300 UV254 Polyester Sheets
Shop All Macherey-Nagel ProductsDescription
TLC Polyester sheets, cellulose layer, POLYGRAM CEL 300 UV254
- Base material: Polyester sheets coated in cellulose
- Mode: Normal Phase
- Layer Hardness: Standard, Soft
Adsorbent:
- Polyester sheets coated in cellulose
- Phase CEL 300
- UV Indicator: Yes
- Particle Type: Fully porous particles (FPP)
- Particle Shape: Irregular
- Particle size 2-20 μm
- Binder: Yes
Indicators:
This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).
Binder:
Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Recommended Applications:
Amino acids, Carbohydrates, Carboxylic acids, Polar substances
General TLC Plate Features from Macherey-Nagel:
- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- The plate can be easily labelled for clear identification
- Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
- Increased separation efficiency due to an optimized particle size distribution
- A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
Specifications
Specifications
| Quantity | 2 Pk. |
| Type | Polyester Sheet |
| Fluorescent Indicator | UV254 |
| Thickness (Metric) | 0.1 mm |
| Dimensions (L x W) | 5 x 20 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.