missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram CEL 300 UV254 Polyester Sheets

Product Code. 10565571 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 10565571

Brand: Macherey-Nagel™ 801023

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

TLC polyester sheets with native fibrous cellulose. Fiber length (95 %) 2–20 μm, average degree of polymerization 400–500, specific surface acc. to Blaine 15000 cm2/g; ≤ 20 ppm Fe, 6 ppm Cu, 7 ppm P; dichloromethane extract ≤ 0.25 %; residue on ignition at 850 °C ≤ 1500 ppm.

TLC Polyester sheets, cellulose layer, POLYGRAM CEL 300 UV254

  • Base material: Polyester sheets coated in cellulose
  • Mode: Normal Phase
  • Layer Hardness: Standard, Soft

Adsorbent:

  • Polyester sheets coated in cellulose
  • Phase CEL 300
  • UV Indicator: Yes
  • Particle Type: Fully porous particles (FPP)
  • Particle Shape: Irregular
  • Particle size 2-20 μm
  • Binder: Yes

Indicators:

This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).

Binder:

Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Recommended Applications:

Amino acids, Carbohydrates, Carboxylic acids, Polar substances

 

General TLC Plate Features from Macherey-Nagel:

  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
  • The plate can be easily labelled for clear identification
  • Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
  • Increased separation efficiency due to an optimized particle size distribution
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber

Specifications

Quantity 25 Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Thickness (Metric) 0.1 mm
Dimensions (L x W) 20 x 20 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.