missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygram IONEX-25 SB-AC Polyester Sheets
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Specifications
Specifications
| Quantity | 25 Pk. |
| Type | Polyester Sheet |
| Thickness (Metric) | 0.2 mm |
| Dimensions (L x W) | 20 x 20 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction