missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ SIL G-25 / UV254 Glass Plates

Product Code. 12827951 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
2.5 x 7.5 cm
5 x 10 cm
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12827951 5 x 10 cm Pack of 200
12810951 2.5 x 7.5 cm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12827951 Supplier Macherey-Nagel™ Supplier No. 809027.200
  • ONLINE EXCLUSIVE PRICE
    368.49 €
    337.05 € / Pack of 200
    Save 31.44 €
    8% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stable in almost all organic solvents. Macherey-Nagel™ SIL G-25 / UV254 Glass Plates are analytical plates with unmodified standard silica layers on glass for use in TLC. Feature a green fluorescence indicator for short-wave UV.

SIL G-25 / UV254 Glass Plates feature unmodified, standard-grade silica 60 layers with a green fluorescent indicator.

  • Manganese-activated zinc silicate indicator with green fluorescence for short-wave UV (254nm)
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualization reagents

Specifications

Material Glass Plates with Unmodified Silica 60 Layers
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 200 Pk.
Type Glass TLC Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.25 mm
Dimensions (L x W) 5 x 10 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.