missing translation for 'onlineSavingsMsg'
Learn More

Polyimide film, 0.008mm thick, Thermo Scientific Chemicals

Product Code. 11344867
Change view
Click to view available options
Quantity:
100 x 100 mm
150 x 150 mm
300 x 300 mm
Unit Size:
100mm
150mm
300mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344867 100 x 100 mm 100mm
11334867 150 x 150 mm 150mm
11354867 300 x 300 mm 300mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11344867 Supplier Thermo Scientific Alfa Aesar Supplier No. 041896.GH
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 62929-02-6
Molecular Formula C35H28N2O7
Molecular Weight (g/mol) 588.62
MDL Number MFCD00677744
InChI Key CDTDIQLZIBORMV-UHFFFAOYNA-N
Synonym polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione
PubChem CID 6454485
IUPAC Name 1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
SMILES CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1

Especificaciones

Physical Form ≈0.1136g/100x100mm
Quantity 100 x 100 mm
Chemical Name or Material Polyimide film, 0.008mm thick

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.