missing translation for 'onlineSavingsMsg'
Learn More

Polyimide film, 0.025mm thick x 20mm width, Thermo Scientific Chemicals

Product Code. 15411737
Change view
Click to view available options
Quantity:
33 m
Unit Size:
33m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411737 33 m 33m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15411737 Supplier Thermo Scientific Alfa Aesar Supplier No. 041898.UL
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62929-02-6
Physical Form Film
Molecular Formula C35H28N2O7
MDL Number MFCD00677744
Synonym polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione
Quantity 33 m
InChI Key CDTDIQLZIBORMV-UHFFFAOYNA-N
SMILES CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1
IUPAC Name 1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 588.62
PubChem CID 6454485
Chemical Name or Material Polyimide, 0.025mm thick x 20mm width
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.