Learn More
Thermo Scientific™ Polymer Tubing Cutter
Product Code. 10344573 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Replacement Blade
Tubing Cutter
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10344573 Tubing Cutter Each
10759295 Replacement Blade Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10344573 Supplier Thermo Scientific™ Supplier No. A327
Estimated Shipment: 09-12-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using Thermo Scientific™ Polymer Tubing Cutter.
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using the Thermo Scientific™ Polymer Tubing Cutter. It is compatible with rigid polymeric tubing and comes with guide holes for 1/16 in. and 1/8 in. tubing.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Cutter
Description 90° Burr-Free End
Quantity 1 Each
For Use With (Equipment) Tubing
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.