missing translation for 'onlineSavingsMsg'
Learn More

Polyoxyethylene bis(amine), M.W. 8,000

Product Code. 15462797
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462797 250 mg 250mg
15442797 1 g 1g
15452797 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462797 Supplier Thermo Scientific Alfa Aesar Supplier No. 046903.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24991-53-5 | H2NCH2CH2(OCH2CH2)nNH2

It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24991-53-5
Molecular Formula H2NCH2CH2(OCH2CH2)nNH2
MDL Number MFCD00132004
Synonym PEG-diamine

Specifications

Color White
Physical Form Granular Solid
Solubility Information It is soluble in organic solvents. It is insoluble in water.
Quantity 250 mg
Sensitivity Air Sensitive
Chemical Name or Material Polyoxyethylene bis (amine), M.W. 8,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.