missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips

Product Code. 10418212
Change view
Click to view available options
Color:
Blue
Green
Red
Turquoise
Yellow
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10418212 Blue Pack of 10
10458782 Green Pack of 10
10204241 Red Pack of 10
10244141 Turquoise Pack of 10
10579941 Yellow Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10418212 Supplier Lenz Laborglasinstrumente™ Supplier No. 01103719
  • ONLINE EXCLUSIVE PRICE
    54.40 €
    35.97 € / Pack of 10
    Save 18.43 €
    33% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensures a secure connection while preventing damage to glass tubing and increasing safety in the laboratory. Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips are manufactured from temperature resistant polyoxymethylene (POM).

  • Good spring properties
  • Color coded by size
  • Will not corrode or bend out of shape

Specifications

Quantity 10/Pk.
Type Joint Clip
Material POM
For Use With (Equipment) For conical ground joints
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.