missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric acid, ≥83% phosphate (as P2O5) basis, Solstice

Product Code. 15690700 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15690700 500 g 500g
15670700 1 kg 1kg
15680700 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15690700 Supplier Honeywell-Fluka Supplier No. 04101500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

CAS Max % ≤100%
Quantity 500 g
Percent Purity >83%
Assay of phosphate as P2O5
Chemical Name or Material Polyphosphoric Acid, phosphate (as P2O5) ≥83%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.