Learn More
Chemsplash Polyprop Dust Suit Coverall, Zip Front Navy
Product Code. 30058417
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058417 3X-Large Pack of 50
30058395 Small Pack of 50
30058399 Medium Pack of 50
30058342 Large Pack of 50
30058213 X-Large Pack of 50
30058303 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058417 Supplier Chemsplash Supplier No. 2523.300.009
  • ONLINE EXCLUSIVE PRICE
    121.41 €
    112.73 € / Pack of 50
    Save 8.68 €
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Polypropylene Dust Suit Coverall is made from a 30gsm non-woven material and comes with a full zip front closure, elasticated wrist and ankle to secure fit and has a hood.
Polypropylene Dust Suit Coverall
  • 30GSM Non-Woven Polypropylene
  • Zip Front
  • Elasticated Wrist/Ankle
  • Hood
  • Suitable for Hygiene Rules / Food Processing
  • Clean Areas / Disposal Operations
  • PFAS Free
Description: The Polypropylene Dust Suit Coverall is made from a 30GSM non-woven material and features a full zip front closure, elasticated wrist and ankle for a secure fit, and a hood. Individually packed for hygiene purposes, it is suitable for Hygiene Rules, Food Processing, Clean Areas, and Disposal Operations.
Specifications
Product Type Hooded Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None