Learn More
Chemsplash Polyprop Dust Suit Coverall, Zip Front Navy
GREENER_CHOICE
Product Code. p-200105876
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Product Code. 30058395
Brand: Chemsplash 2523.300.004
Only null left
Add to basket
Add to basket
Polypropylene Dust Suit Coverall is made from a 30gsm non-woven material and comes with a full zip front closure, elasticated wrist and ankle to secure fit and has a hood.
Polypropylene Dust Suit Coverall
  • 30GSM Non-Woven Polypropylene
  • Zip Front
  • Elasticated Wrist/Ankle
  • Hood
  • Suitable for Hygiene Rules / Food Processing
  • Clean Areas / Disposal Operations
  • PFAS Free
Description: The Polypropylene Dust Suit Coverall is made from a 30GSM non-woven material and features a full zip front closure, elasticated wrist and ankle for a secure fit, and a hood. Individually packed for hygiene purposes, it is suitable for Hygiene Rules, Food Processing, Clean Areas, and Disposal Operations.
Specifications
Product Type Hooded Coveralls
Size Small
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None