missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ A71 Chemical Spray Protection Coveralls

Product Code. 12641396
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12641396

Brand: KleenGuard™ 96780

This item is not returnable. View return policy

Coveralls create a spray-tight and liquid-tight barrier against harmful substances when working with hazardous chemicals.

  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
  • Highly visible yellow color - for improved worker safety
  • Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
  • Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
  • Suitable for use as protection against Infective Agents and Biohazards
  • Suitable for use as protection against solid particles and particulate radioactive contamination
  • Treated to be antistatic
  • EN 1073-2 approved for radioactive dust protection
  • Anti-static EN 1149-1 for critical areas

Specifications

Product Type General Purpose Coveralls
Size X-Large
Material Spunbond Polypropylene
Color Yellow
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class Type 3, 4 and 5
Resistance Perforation, Flame, Abrasion, Tears, Particle, Liquid
Latex Free Yes
No. of Pockets None
For Use With (Application) Hazardous Chemicals
Style Full Body
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.