missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Adapter Support
Shop All Brand Products
Click to view available options
Color:
Dark Blue
Green
Magenta
Unit Size:
Each
Description
- Constructed of polypropylene
- For use with accu-jet™
Specifications
Specifications
| Color | Magenta |
| Type | Adapter Support |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction