missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Polypropylene Bouffant Cap in Polybag, 48 cm Diameter

Product Code. 17720129 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Green
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17720129 Blue Pack of 100
17710129 White Pack of 100
17730129 Green Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17720129 Supplier Kolmi Hopen™ Supplier No. 70.002
  • ONLINE EXCLUSIVE PRICE
    5.33 €
    3.67 € / Pack of 100
    Save 1.66 €
    31% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These comfortable bouffant caps provide additional hygiene control for various industrial environments.

  • Lightweight polypropylene cap improves user's comfort
  • Ability to use a color code for tracking
  • Adapts to different head sizes

Specifications

Material Polypropylene
Color Blue
Disposable Single-use
Design Type Bouffant
Sterility Non-sterile
Certifications/Compliance EN ISO 9001
Packaging Type Polybag
Quantity 100 Pk.
Length (Metric) 48 cm
Diameter (Metric) 48 cm
Construction Sewn and Elasticated
For Use With (Application) HACCP Processes and Various Industries
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.