Learn More
BRAND™ Polypropylene Burette Clamps
To be mounted on support rods of 8 to 14mm dia. Easily affixes the burette by clamping with a non-corroding spring.
Brand:  BRAND™ 16505
Additional Details : Weight : 0.04500kg
 View more versions of this product
Product Code. 10017830
  • 47.15 € / Each
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Polypropylene construction
  • White
  • For use with support rods 8 to 14mm dia.
Specifications
Specifications
White
Polypropylene
Burette Holder
For use with 1 Burette
1 Each