missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Cap Inserts

Product Code. 10106390 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10106390 White Pack of 500
10462321 Blue Pack of 500
10054680 Yellow Pack of 500
10648271 Green Pack of 500
10771791 Red Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10106390 Supplier BRAND™ Supplier No. 114850
  • ONLINE EXCLUSIVE PRICE
    19.23 €
    17.78 € / Pack of 500
    Save 1.45 €
    7% Off
View all promotions
In Stock
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Providing a secure fit for all sizes.

  • Available in different colors
  • Provided in packs of 500
  • High temperature and chemical resistance

TRUSTED_SUSTAINABILITY

Specifikationer

Color White
Material Polypropylene
Quantity 500/Pk.
For Use With (Equipment) Fits All Sizes
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.