missing translation for 'onlineSavingsMsg'
Learn More

CellPath Cellstor Pot Pre-filled Specimen Container

Product Code. 13191184
Change view
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13191184 60 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13191184 Supplier CellPath Supplier No. BAF600008A
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Improve laboratory speed and efficiency by using specimen containers pre-filled with neutral buffered formalin. Cellpath™ Cellstor Pot Pre-filled Specimen Container uses an innovative sealing mechanism to ensure a reliable, leak-proof seal.

  • Leak-tight lid ensures safe transport and storage of specimens.
  • Packed in trays for easy handling.
  • Pre-filled with buffered formalin: reduces your exposure to formalin fumes.
  • Batch numbered for full traceability.
  • Range of sizes available.

Specifications

Product Type Pre-filled Specimen Container
Material Polypropylene
Capacity (Metric) 60 mL
Includes 10% neutral buffered formalin
Label Yes
No. per Pack 25/Pk.
compliance-icons compliance-icons
Product Identifier
  • 10% Neutral Buffered Formalin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: formaldehyde, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.