missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Clear Autoclave Bags

Product Code. 16207971 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
310 x 660 mm
410 x 630 mm
620 x 780 mm
Model:
Biohazard Waste Identification
Plain without Print
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16207971

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

  • For high temperature sterilisation (134°C) to a maximum of 140°C
  • 40μm thickness
  • High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
  • For safe disposal of biohazard waste
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Printed with a biohazard warning

Specifications

Color Clear
Dimensions (L x W) 620 x 780 mm
Model Biohazard Waste Identification
Quantity 100/Pk.
Width (Metric) 620 mm
Autoclavable Autoclavable
Includes Biohazard Warning
Thickness 40 μm
Material Polypropylene
Temperature (Metric) 134°C to 140°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.