missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Clear Autoclave Bags

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

Brand:  Fisherbrand™

Additional Details : Weight : 1.46700kg

 View more versions of this product

Product Code. 16217971

  • 27.24 € / Pack of 100
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

  • For high temperature sterilisation (134°C) to a maximum of 140°C
  • 40μm thickness
  • High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
  • For safe disposal of biohazard waste
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Printed with a biohazard warning
Specifications

Specifications

Plain translucent bag, no print
Clear
40 μm
310 x 660 mm
310 mm
Autoclavable
Polypropylene
100/Pk.
134°C to 140°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers