missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Clear Autoclave Bags
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
310 x 660 mm
410 x 630 mm
620 x 780 mm
Model:
Biohazard Waste Identification
Plain without Print
missing translation for 'unitSize'
Pack of 100
Beschreibung
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
- For high temperature sterilisation (134°C) to a maximum of 140°C
- 40μm thickness
- High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
- For safe disposal of biohazard waste
- Specifically designed for contaminated waste disposal in autoclaves or incinerators
- Printed with a biohazard warning
Spezifikation
Spezifikation
| Color | Clear |
| Dimensions (L x W) | 410 x 630 mm |
| Height (Metric) | 630 mm |
| Model | Plain without Print |
| Quantity | 100/Pk. |
| Width (Metric) | 410 mm |
| Autoclavable | Autoclavable |
| Thickness | 40 μm |
| Material | Polypropylene |
| Temperature (Metric) | 134°C to 140°C |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur