missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene Cleats For 120/277V Lockout

Product Code. 11751346
Click to view available options
Color:
Red
Unit Size:
1 pack
This item is not returnable. View return policy

Product Code. 11751346

Brand: Brady™ undefined

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Cleats
Alternate Part Number LOCKOUT CLEAT 277 VOLT GENERAL USE PK6
Color Red
Description Extra Cleats for 120/277V Lockout
For Use With (Equipment) For 120/277V Lockout
Quantity 6 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.