missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polypropylene Compressed Air Magnetic Stirrer

Product Code. 11897933 Shop All Cole-Parmer Products
Click to view available options
Capacity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11897933

Brand: Cole-Parmer™ 0463400

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Corrosion-proof plastic eliminates shock hazard in dangerous environments.

  • Can be used in hazardous locations or on your benchtop
  • Position stirrer off your benchtop by connecting it to a 1/2inch or 3/8inch dia support rod with the built-in ring clamp
  • Inlet ID measures 4.1 mm; outlet ID measures 9.1 mm
  • Attach stirrer to any air pressure line or water source - stirs 500mL of water at 800rpm with air at 8psi or with water at 12psi
  • Includes: 6ft of vinyl inlet and 1ft outlet tubing for easy hookup to your air or water source

Specifications

Speed 0 to 800 rpm
Stirring Positions 1
Top Plate Material Polypropylene
Capacity 500 mL
Diameter (Metric) 127 mm
Diameter (Metric) Outside 9.1 mm
Length (Metric) 127 mm
Top Plate Diameter (Metric) 127 mm
Includes 6-ft of Vinyl Inlet and 1-ft Outlet Tubing For Easy Hookup To Air or Water Source
Type Stirrer
Weight (Metric) 0.8 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.