Learn More
Fisherbrand™ Polypropylene Crystal Pipet Tip E
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
Unit Size:
Pack of 1000
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
Specifications
Specifications
| Volume (Metric) | 0.5 to 10 μL |
| Color | Crystal |
| Autoclavable | Autoclavable |
| Material | Polypropylene |