Learn More
Eppendorf™ Polypropylene DNA LoBind Polypropylene Microcentrifuge Tube
Maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding with these tubes. Eppendorf™ Polypropylene DNA LoBind Tubes are ideal for sample preparation and long-term storage.
Brand: Eppendorf™ 0030108078
17.98 EUR valid until 2023-06-30
Use promo code "19137" to get your promotional price.
Code : 96
Additional Details : Weight : 0.36000kg
Description
Maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding with these tubes. Eppendorf™ Polypropylene DNA LoBind Tubes are ideal for sample preparation and long-term storage.
- Free from DNA, DNase, RNase and ATP (PCR* clean)
- Less than 1% loss of DNA /RNA at low concentrations and critical conditions (high salt, short DNA-fragments)
- Special PP type manufactured under optimized conditions
- Available as 0.5mL, 1.5mL and 2mL Safe-Lock™ tubes
- High degree of transparency
Specifications
Polypropylene | |
Transparent | |
30000xG | |
250 Tubes (5 bags of 50) | |
Yes |
2mL | |
Polypropylene | |
DNA LoBind consumables are perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology | |
30000xG |