missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene Glycol 2000 Grade Extra Pure, SLR, Fisher Chemical

Product Code. 10764761 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10764761 1 L Amber glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10764761 Supplier Fisher Chemical Supplier No. P/3694/15
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 25322-69-4
Physical Form Viscous liquid Liquid
Molecular Formula (C3H6O)n
MDL Number MFCD00084446
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
Quantity 1 L
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES [H]OC(C)CO
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
Molecular Weight (g/mol) 134.18
PubChem CID 32881
Packaging Amber glass bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.