missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene glycol 2000

Product Code. 11343646
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343646 100 mL 100mL
11353646 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343646 Supplier Thermo Scientific Alfa Aesar Supplier No. L14699.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

Used to make polyester compounds, absorb extra water and maintain moisture in certain medicines, cosmetics, or food products and as a base for deicing solutions. Propylene glycol is used by the chemical, food, and pharmaceutical industries as an antifreeze. It is a solvent for food colors and flavors, and in the paint and plastics industries. Propylene glycol is also used to create artificial smoke or fog used in fire-fighting training and in theatrical productions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Density 1.002
Solubility Information Miscible with water,ethanol,diethyl ether and chloroform.
Quantity 100 mL
Formula Weight 2000
Flash Point 194°C (381°F)
Refractive Index 1.451
Chemical Name or Material Polypropylene glycol 2000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.