missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene Glycol (P 400), TRC

Product Code. 30405910
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405910 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405910 Supplier Toronto Research Chemicals Supplier No. TRCH544090100ML
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polypropylene Glycol (P 400)
CAS 25322-69-4
Molecular Formula (C3H6O)nH2O
Quantity 100 mL
Synonym 2-(2-Hydroxypropoxy)propan-1-ol
InChI Formula InChI=1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3
SMILES CC(O)COC(C)CO
Molecular Weight (g/mol) 76.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.