missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Grid Inserts

Brand:  Ratiolab™ 5220100

Code : A0

Additional Details : Weight : 0.03420kg

 View more versions of this product

Product Code. 10022482

  • 14.00 € / Pack of 5

Estimated Shipment: 15-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Polypropylene
Insert Grid
5 Pack
133 mm
Square
20.5 mm Diameter Tubes
133 mm
30 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers