missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Hexagonal Key

Product Code. 10395182 Shop All Bürkle Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10395182 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10395182 Supplier Buerkle™ Supplier No. 06000001
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters. The Polypropylene Hexagonal Key helps to connect.

  • Hexagonal Key for screwing adapters
  • Made of Polypropylene
  • For LaboPlast™ thread adapters
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hex Key
For Use With (Equipment) Thread Adapter
Material Polypropylene
Color Blue
No. per Pack 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.