Learn More
Chemsplash Polypropylene Lab/Visitors Coat Navy with Velcro Closure
GREENER_CHOICE
Product Code. 30058163
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Product Code. 30058163
Brand: Chemsplash 2515.200.005
Only null left
Add to basket
Add to basket
Visitors Polypropylene Disposable Lab Coat with a velcro front closure. Suitable for visitors, site inspections or general day to day wear in minimal risk environments.
Visitors Polypropylene Disposable Lab Coat
  • Material: 30gsm Polypropylene Fabric
  • Features:
    • Velcro Closure
    • With Fold Over Collar
    • Elasticated Cuffs
    • Ideal for Non-Hazardous Environments
    • Ideal for Food Industry
    • PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Specifications
Product Type Visitor Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color Navy
Closure Type Hook and Loop
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
Style Knee-length
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable